2-Ethoxyethanol ≥99% for spectrophotometry

Danger

256374-2L 256374-1L
220048-915EA 105.85 USD
220048-915 220048-914
2-Ethoxyethanol ≥99% for spectrophotometry
2-Ethoxyethanol
Formula: C₂H₅OCH₂CH₂OH
Boiling Pt: 135 °C (1013 hPa)
Melting Pt: –70 °C
Density: 0.932 g/cm³ (20 °C)
Flash Pt: 40 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002869
CAS Number: 110-80-5
EINECS: 203-804-1
UN: 1171
ADR: 3,III
REACH: 01-2119560582-38
Merck Index: 13,03786
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=461a4daf8709ebd6b4516d73b4b32eee33cd1bca&vid=625adefb8d1e2126363931c57a51d3e1df419926&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR