2-Butoxyethanol ≥99.0% for spectrophotometry

Warning

Synonyms: Ethylene glycol butyl ether, Butyl cellosolve, Ethylene glycol monobutyl ether

256366-1L 256366-2L
220048-912EA 46.15 USD
220048-912 220048-913
2-Butoxyethanol ≥99.0% for spectrophotometry
2-Butoxyethanol
Formula: CH₃(CH₂)₃OCH₂CH₂OH
MW: 118.18 g/mol
Boiling Pt: 171 °C (1013 hPa)
Melting Pt: -70 °C
Density: 0.902 g/cm³ (20 °C)
Flash Pt: 61 °C
Storage Temperature: Ambient
MDL Number: MFCD00002884
CAS Number: 111-76-2
EINECS: 203-905-0
UN: 2810
ADR: 6.1,III
Merck Index: 13,01558
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba5a9d2c527d0c167d2248d1936e2d1bd4aeab7a&vid=75260b2608ddc5eb29642178ab193fbcaf54198d&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR