Lithium carbonate ≥99.0% ACS

Warning

255823-500G 255823-100G
220047-583EA 109.54 USD
220047-583 220047-582
Lithium carbonate ≥99.0% ACS
Lithium carbonate
Formula: Li₂CO₃
Melting Pt: 720 °C
Density: 2.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011084
CAS Number: 554-13-2
EINECS: 209-062-5
Merck Index: 13,05549

Order Now

Specification Test Results

Assay 99.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR