Magnesium chloride hexahydrate ≥99.995%

255777-25G 255777-5G 255777-100G
220048-720EA 398.46 USD
220048-720 220048-721 220048-719
Magnesium chloride hexahydrate ≥99.995%
Magnesium chloride hexahydrate
Formula: MgCl₂·6H₂O
Boiling Pt: 1412 °C (1013 hPa)
Melting Pt: 117 °C
Density: 1.57 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149781
CAS Number: 7791-18-6
EINECS: 232-094-6
Merck Index: 13,05684

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR