Cesium fluoride ≥99.99%

Danger

255718-50G 255718-10G
220048-714EA 1004.62 USD
220048-714 220048-713
Cesium fluoride ≥99.99%
Cesium fluoride
Formula: CsF
MW: 151.9 g/mol
Boiling Pt: 1251 °C (1013 hPa)
Melting Pt: 680 °C
Density: 4.627 g/cm³ (20 °C)
MDL Number: MFCD00010960
CAS Number: 13400-13-0
EINECS: 236-487-3
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR