Hydroxylammonium chloride ACS

Warning

Synonyms: Hydroxylamine HCl, Hydroxylamine hydrochloride

255580-500G 255580-100G 255580-5G
220048-697EA 129.85 USD
220048-697 220048-696 220048-698
Hydroxylammonium chloride ACS
Hydroxylammonium chloride
Formula: NH₂OH·HCl
MW: 69.49 g/mol
Boiling Pt: 306 °C (1013 hPa)
Melting Pt: 151 °C
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00051089
CAS Number: 5470-11-1
EINECS: 226-798-2
UN: 2923
ADR: 8,III
Merck Index: 13,04853

Order Now

Specification Test Results

Assay 98.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR