Potassium chlorate ≥99.0% ACS

Danger

255572-100G 255572-500G
220048-694EA 45.54 USD
220048-694 220048-695
Potassium chlorate ≥99.0% ACS
Potassium chlorate
Formula: KClO₃
Boiling Pt: 400 °C (1013 hPa)
Melting Pt: 356 °C
Density: 2.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011361
CAS Number: 3811-04-9
EINECS: 223-289-7
UN: 1485
ADR: 5.1,II
Merck Index: 13,07703

Order Now

Specification Test Results

Assay 99.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR