3-Bromobiphenyl (PBB No. 2) ≥97%

Synonyms: PBB No. 2

255386-1G 255386-5G
220048-674EA 26.92 USD
220048-674 220048-675
3-Bromobiphenyl (PBB No. 2) ≥97%
3-Bromobiphenyl (PBB No. 2)
Formula: C₁₂H₉Br
MW: 233.11 g/mol
Boiling Pt: 299 °C (1013 hPa)
Melting Pt: 9 °C
Density: 1.41 g/cm³ (20 °C)
MDL Number: MFCD00000082
CAS Number: 2113-57-7
EINECS: 218-304-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR