2-Thiophenesulfonyl chloride

Danger

Synonyms: Thiophene-2-sulphonyl chloride

255335-5G 255335-1G 255335-25G
220048-672EA 51.48 USD
220048-672 220048-670 220048-671
2-Thiophenesulfonyl chloride
2-Thiophenesulphonyl chloride
Formula: C₄H₃ClO₂S₂
MW: 182.65 g/mol
Boiling Pt: 82…84 °C (1 mmHg)
Melting Pt: 32 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005426
CAS Number: 16629-19-9
EINECS: 240-677-1
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR