3-Aminosalicylic acid ≥97%

Warning

255300-100MG 255300-500MG
220048-666EA 100.15 USD
220048-666 220048-667
3-Aminosalicylic acid ≥97%
3-Aminosalicylic acid
Formula: C₇H₇NO₃
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 290 °C
Storage Temperature: Ambient
MDL Number: MFCD00010299
CAS Number: 570-23-0
EINECS: 209-328-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR