α'-Bromo-α,α,α-trifluoro-o-xylene ≥96%

Danger

254967-5G 254967-1G
220048-630EA 42.54 USD
220048-630 220048-629
α'-Bromo-α,α,α-trifluoro-o-xylene ≥96%
α'-Bromo-α,α,α-trifluoro-o-xylene
Formula: C₈H₆BrF₃
Boiling Pt: 116…118 °C (60 mmHg)
Melting Pt: 30 °C
Density: 1.57 g/cm³ (20 °C)
MDL Number: MFCD00013558
CAS Number: 395-44-8
EINECS: 206-901-7
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR