2,3-Dichlorobenzaldehyde ≥99%

Warning

254835-25G 254835-100G
220048-620EA 44.37 USD
220048-620 220048-619
2,3-Dichlorobenzaldehyde ≥99%
2,3-Dichlorobenzaldehyde
Formula: C₇H₄Cl₂O
MW: 175.01 g/mol
Boiling Pt: 243 °C (1013 hPa)
Melting Pt: 64 °C
Density: 1.33 g/cm³ (100 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010127
CAS Number: 6334-18-5
EINECS: 228-711-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR