Triisopropanolamine ≥95%

Synonyms: 1,1',1''-Nitrilotripropan-2-ol, 1,1',1"-Nitrilotri-2-propanol, Tris(2-hydroxypropyl)amine

254746-100G 254746-1KG
220048-610EA 50.22 USD
220048-610 220048-611
Triisopropanolamine ≥95%
Triisopropanolamine
Formula: C₉H₂₁NO₃
MW: 191.27 g/mol
MDL Number: MFCD00004533
CAS Number: 122-20-3
EINECS: 204-528-4

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR