di-Lithium tetraborate ≥99.995% (trace metals basis), Sigma-Aldrich®

Synonyms: Lithium tetraborate, Dilithium tetraborate

254304-25G 254304-100G
220048-521EA 118.46 USD
220048-521 220048-520
di-Lithium tetraborate ≥99.995% (trace metals basis), Sigma-Aldrich®
di-Lithium tetraborate
Formula: Li₂B₄O₇
MW: 169.12 g/mol
Melting Pt: 918 °C
Density: 1.4 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011083
CAS Number: 12007-60-2
EINECS: 234-514-3
Merck Index: 14,05524

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR