Lithium hydroxide monohydrate ≥99.95%

Danger

254274-250G 254274-50G 254274-10G
220048-111EA 1015.38 USD
220048-111 220048-112 220048-110
Lithium hydroxide monohydrate ≥99.95%
Lithium hydroxide monohydrate
Formula: LiOH·1H₂O
MW: 41.96 g/mol
Boiling Pt: 920 °C (1013 hPa)
Melting Pt: 462 °C
Density: 1.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149772
CAS Number: 1310-66-3
EINECS: 215-183-4
UN: 2680
ADR: 8,II
Merck Index: 13,05556

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR