Iron(III) nitrate nonahydrate

Warning

Synonyms: Ferric nitrate nonahydrate, Iron trinitrate nonahydrate, Iron(III) nitrate hydrate, Ferric Nitrate, 9-Hydrate

254223-10G 254223-250G 254223-50G
220048-101EA 74.77 USD
220048-101 220048-102 220048-103
Iron(III) nitrate nonahydrate
Iron(III) nitrate nonahydrate
Formula: Fe(NO₃)₃·9H₂O
MW: 404 g/mol
Boiling Pt: 125 °C (1013 hPa)
Melting Pt: 47 °C
Density: 1.68 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149708
CAS Number: 7782-61-8
EINECS: 233-899-5
UN: 1466
ADR: 5.1,III
Merck Index: 13,04057

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR