Ammonium chloride

Warning

254134-100G 254134-25G 254134-5G
220048-509EA 467.69 USD
220048-509 220048-510 220048-511
Ammonium chloride
Ammonium chloride
Formula: NH₄Cl
Melting Pt: 335 °C
Density: 1.5256 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011420
CAS Number: 12125-02-9
EINECS: 235-186-4
Merck Index: 13,00510
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e45c4909ae60875d5b9dca2f28f54f5c8028e64&vid=0a37e08e2cb01fc6e0bca4804b48ba22a9054eb3&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR