Europium trinitrate hydrate ≥99.99%

Synonyms: Europium (III) nitrate hydrate

254061-1G 254061-10G
220048-500EA 75.08 USD
220048-500 220048-499
Europium trinitrate hydrate ≥99.99%
Europium trinitrate hydrate
Formula: EuN₃O₉·xH₂O
MW: 337.98 g/mol (anhy)
MDL Number: MFCD00149698
CAS Number: 100587-95-9
EINECS: 233-380-3
UN: 1477
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR