Pyrogallol ≥99% ACS

Warning

Synonyms: Benzene-1,2,3-triol, 1,2,3-Trihydroxybenzene

254002-10G 254002-250G 254002-50G
220048-488EA 23.85 USD
220048-488 220048-489 220048-490
Pyrogallol ≥99% ACS
Pyrogallol
Formula: C₆H₆O₃
MW: 126.11 g/mol
Boiling Pt: 309 °C (1013 hPa)
Melting Pt: 130 - 132 °C
Density: 1.453 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002192
CAS Number: 87-66-1
EINECS: 201-762-9
UN: 2811
ADR: 6.1,III
Merck Index: 13,08090
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09c5271be2848ebfa6e3614bff56b329a59cb6fe&vid=db25c87d92a56d9ff5d36f04d6bbf59acf236659&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR