4-tert-Butyl-1-bromobenzene ≥97%

Synonyms: 1-Bromo-4-tert-butylbenzene, 2-(4-Bromophenyl)-2-methylpropane, 1-Bromo-4-(tert-butyl)benzene, 4-Bromo-tert-butylbenzene

253928-100G 253928-25G
220048-482EA 133.54 USD
220048-482 220048-483
4-tert-Butyl-1-bromobenzene ≥97%
4-tert-Butyl-1-bromobenzene
Formula: C₁₀H₁₃Br
MW: 213.12 g/mol
Boiling Pt: 232 °C (1013 hPa)
Melting Pt: 15.5 °C
Density: 1.2572 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000108
CAS Number: 3972-65-4
EINECS: 223-599-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR