(±)-3-Quinuclidinol

Synonyms: (±)-Quinuclidin-3-ol, (±)-3-Hydroxyquinuclidine

253340-25G 253340-5G
220047-817EA 151.85 USD
220047-817 220047-818
(±)-3-Quinuclidinol
(±)-3-Quinuclidinol
Formula: C₇H₁₃NO
MW: 127.19 g/mol
Melting Pt: 225…227 °C
MDL Number: MFCD00151326
CAS Number: 1619-34-7
EINECS: 216-578-4
UN: 3263
ADR: 8,III
Merck Index: 12,08266

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR