Tris(diethylamino)phosphine ≥97%

Synonyms: Hexaethyltriaminophosphine, Hexaethylphosphorus triamide, Hexaethyl phosphorous triamide

253189-25G 253189-5G
220048-430EA 128.77 USD
220048-430 220048-431
Tris(diethylamino)phosphine ≥97%
Tris(diethylamino)phosphine
Formula: C₁₂H₃₀N₃P
MW: 247.36 g/mol
Density: 0.89 g/cm³ (20 °C)
MDL Number: MFCD00009041
CAS Number: 2283-11-6
EINECS: 218-920-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR