Methoxytrimethylsilane

Synonyms: Trimethyl(methoxy)silane

253006-100G 253006-250G 253006-25G
220047-989EA 370.08 USD
220047-989 220048-403 220048-404
Methoxytrimethylsilane
Methoxytrimethylsilane
Formula: C₄H₁₂OSi
MW: 104.22 g/mol
Boiling Pt: 49…51 °C (1013 hPa)
Density: 0.755 g/cm³ (20 °C)
MDL Number: MFCD00008344
CAS Number: 1825-61-2
EINECS: 217-369-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR