Diphenylphosphine

252964-10G 252964-50G
220047-980EA 57.05 USD
220047-980 220047-981
Diphenylphosphine
Diphenylphosphine
Formula: C₁₂H₁₁P
MW: 186.19 g/mol
MDL Number: MFCD00003040
CAS Number: 829-85-6
EINECS: 212-591-4
UN: 2845
ADR: 4.2,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR