Tropone

Synonyms: Cyclohepta-2,4,6-trien-1-one, 2,4,6-Cycloheptatrienone, 2,4,6-Cycloheptatrien-1-one

252832-1G 252832-5G
220048-399EA 89.28 USD
220048-399 220048-400
Tropone
Tropone
Formula: C₇H₆O
MW: 106.12 g/mol
MDL Number: MFCD00014331
CAS Number: 539-80-0
EINECS: 208-725-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR