Methyl-3-hydroxybenzoate ≥99%

Synonyms: Methyl 3-Hydroxybenzoate, 3-Hydroxybenzoic acid methyl ester

252794-25G 252794-100G
220048-393EA 49.11 USD
220048-393 220048-392
Methyl-3-hydroxybenzoate ≥99%
Methyl-3-hydroxybenzoate
Formula: C₈H₈O₃
MW: 152.15 g/mol
MDL Number: MFCD00002295
CAS Number: 19438-10-9
EINECS: 243-071-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR