2-Iodoanisole ≥98%

Synonyms: 1-Iodo-2-methoxybenzene

252786-25G 252786-100G
220048-391EA 61.06 USD
220048-391 220048-390
2-Iodoanisole ≥98%
2-Iodoanisole
Formula: C₇H₇IO
MW: 234.04 g/mol
Boiling Pt: 238 °C (1013 hPa)
Density: 1.445 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001039
CAS Number: 529-28-2
EINECS: 208-456-4
Merck Index: 13,05050

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR