2-Bromo-1,4-dimethoxybenzene ≥98%

Warning

Synonyms: 1-Bromo-2,5-dimethoxybenzene

252743-25G 252743-5G
220048-384EA 67.95 USD
220048-384 220048-385
2-Bromo-1,4-dimethoxybenzene ≥98%
2-Bromo-1,4-dimethoxybenzene
Formula: C₈H₉BrO₂
MW: 217.06 g/mol
Boiling Pt: 107 °C (0.2 -rr)
Density: 1.445 g/cm³ (15 °C)
MDL Number: MFCD00008355
CAS Number: 25245-34-5
EINECS: 246-756-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR