Trimethylsilyl isocyanate

252646-25G 252646-5G
220048-372EA 267.08 USD
220048-372 220048-373
Trimethylsilyl isocyanate
Trimethylsilyl isocyanate
Formula: C₄H₉NOSi
MW: 115.21 g/mol
MDL Number: MFCD00001993
CAS Number: 1118-02-1
EINECS: 214-256-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR