N-(Bromomethyl)phthalimide

Synonyms: 2-(Bromomethyl)isoindoline-1,3-dione

252611-5G 252611-25G
220048-371EA 67.85 USD
220048-371 220048-370
N-(Bromomethyl)phthalimide
N-(Bromomethyl)phthalimide
Formula: C₉H₆BrNO₂
MW: 240.06 g/mol
Boiling Pt: 410 °C (1013 hPa)
Melting Pt: 148…151 °C
MDL Number: MFCD00005897
CAS Number: 5332-26-3
EINECS: 226-239-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR