D-(+)-Sucrose octaacetate

Synonyms: D-(+)-Saccharose octaacetate, Sucrose octaacetate, D(+)-Sucroseoctaacetat, (2R,3R,4S,5R,6R)-2-(Acetoxymethyl)-6-(((2S,3S,4R,5R)-3,4-diacetoxy-2,5-bis(acetoxymethyl)tetrahydrofuran-2-yl)oxy)tetrahydro-2H-pyran-3,4,5-triyl triacetate

252603-250G 252603-1KG
220048-369EA 66.77 USD
220048-369 220048-368
D-(+)-Sucrose octaacetate
D(+)-Sucrose octaacetate
Formula: C₂₈H₃₈O₁₉
MW: 678.6 g/mol
Melting Pt: 80 - 83 °C
Density: 1.257 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00006623
CAS Number: 126-14-7
EINECS: 204-772-1
Merck Index: 12,09052

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR