8-Quinolinol ≥99% ACS

Warning

Synonyms: 8-Hydroxyquinoline

252565-250G 252565-50G
220048-363EA 113.85 USD
220048-363 220048-364
8-Quinolinol ≥99% ACS
8-Quinolinol
Formula: C₉H₇NO
MW: 145.16 g/mol
Boiling Pt: 267 °C (1013 hPa)
Melting Pt: 71.2 °C
Density: 1.03 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006807
CAS Number: 148-24-3
EINECS: 205-711-1
Merck Index: 13,04869
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=752471e0b5caa5507eb8e74e8054dc401912b3ac&vid=ff81328b844178e10beffb1867c97da0d03eda22&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR