Lactic acid ≥85% in aqueous solution ACS

Danger

Synonyms: DL-Lactic acid, 2-Hydroxypropionic acid, 2-Hydroxypropanoic acid

252476-500G 252476-1KG 252476-100G 252476-2.5KG
220048-350EA 62.62 USD
220048-350 220048-348 220048-347 220048-349
Lactic acid ≥85% in aqueous solution ACS
Lactic acid
Formula: H₃CCH(OH)COOH
MW: 90.08 g/mol
Boiling Pt: 122 °C (20 hPa)
Density: 1.11 to 1.21 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004520
CAS Number: 50-21-5
EINECS: 200-018-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR