Rhodamine 6G

Synonyms: 9-(2-(Ethoxycarbonyl)phenyl)-3,6-bis(ethylamino)-2,7-dimethylxanthylium chloride, 9-[2-(Ethoxycarbonyl)phenyl]-3,6-bis(ethylamino)-2,7-dimethylxanthylium chloride

252433-250MG 252433-1G
220048-344EA 56.62 USD
220048-344 220048-343
Rhodamine 6G
Rhodamine 6G
Formula: C₂₈H₃₁ClN₂O₃
MW: 479.02 g/mol
Boiling Pt: ∼200 °C (1013 hPa)
MDL Number: MFCD00012665
CAS Number: 989-38-8
EINECS: 213-584-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR