Nitrobenzene ≥99.0% ACS

Danger

252379-2L 252379-25ML 252379-500ML
220048-333EA 81.23 USD
220048-333 220047-648 220048-334
Nitrobenzene ≥99.0% ACS
Nitrobenzene
Formula: C₆H₅NO₂
MW: 123.11 g/mol
Boiling Pt: 210.8 °C (1013 hPa)
Melting Pt: 5.7 °C
Density: 1.2037 g/cm³ (20 °C)
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00007043
CAS Number: 98-95-3
EINECS: 202-716-0
UN: 1662
ADR: 6.1,II
Merck Index: 13,06621

Order Now

Specification Test Results

Assay 99.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR