2,6-Dimethylphenyl isocyanate ≥99%

Synonyms: 2,6-Xylyl isocyanate, 2,6-Dimethylphenylisocyanate, 2-Isocyanato-1,3-dimethylbenzene

252336-5G 252336-25G
220047-643EA 82.92 USD
220047-643 220047-642
2,6-Dimethylphenyl isocyanate ≥99%
2,6-Dimethylphenyl isocyanate
Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 87…80 °C (1013 hPa)
Density: 1.057 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002012
CAS Number: 28556-81-2
UN: 2206
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR