8-Bromo-1-octene ≥97%

Warning

252301-1G 252301-5G
220047-639EA 29.94 USD
220047-639 220047-640
8-Bromo-1-octene ≥97%
8-Bromo-1-octene
Formula: C₈H₁₅Br
MW: 191.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000275
CAS Number: 2695-48-9
EINECS: 220-268-4

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR