α-Bromo-2-chlorotoulene ≥97%

Synonyms: 1-(Bromomethyl)-2-chlorobenzene, 2-Chlorobenzyl bromide

252255-5G 252255-25G
220047-633EA 45.23 USD
220047-633 220047-632
α-Bromo-2-chlorotoulene ≥97%
α-Bromo-2-chlorotoulene
Formula: C₇H₆BrCl
MW: 205.48 g/mol
Boiling Pt: 235 °C (1013 hPa)
Density: 1.5897 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000566
CAS Number: 611-17-6
EINECS: 210-257-2
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR