2-Bromobiphenyl (PBB No. 1) ≥96%

Synonyms: 2-Bromobiphenyl, PBB No. 1, 2-Bromodiphenyl

252247-1G 252247-5G
220047-630EA 72.46 USD
220047-630 220047-631
2-Bromobiphenyl (PBB No. 1) ≥96%
2-Bromobiphenyl (PBB No. 1)
Formula: C₁₂H₉Br
MW: 233.11 g/mol
Boiling Pt: 297 °C (1013 hPa)
Melting Pt: 2 °C
Density: 1.2766 g/cm³ (20 °C)
MDL Number: MFCD00000065
CAS Number: 2052-07-5
EINECS: 218-141-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR