1,5-Diiodopentane ≥97%

Warning

Synonyms: Pentamethylene diiodide

252131-100G 252131-25G
220047-621EA 233.69 USD
220047-621 220047-622
1,5-Diiodopentane ≥97%
1,5-Diiodopentane
Formula: C₅H₁₀I₂
MW: 323.94 g/mol
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 9 °C
Density: 2.195 g/cm³ (20 °C)
MDL Number: MFCD00001101
CAS Number: 628-77-3
EINECS: 211-054-1

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR