2,6-Dichlorophenyl isocyanate ≥98%

251879-1G 251879-5G
220046-896EA 263.08 USD
220046-896 220046-897
2,6-Dichlorophenyl isocyanate ≥98%
2,6-Dichlorophenyl isocyanate
Formula: C₇H₃Cl₂NO
MW: 188.01 g/mol
Boiling Pt: 101 °C (5 mmHg)
Melting Pt: 42…44 °C
MDL Number: MFCD00002003
CAS Number: 39920-37-1
EINECS: 254-699-4
UN: 2250
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR