Tetrahydro-1,3-dimethyl-1H-pyrimidin-2-one

Danger

Synonyms: Dimethylpropyleneurea, Dimethyl propylene urea, 1,3-Dimethyl-3,4,5,6-tetrahydro-2(1H)-pyrimidinone, 1,3-Dimethyltetrahydropyrimidin-2(1H)-one, DMPU, 1,3-Dimethyltetrahydro-2(1H)-pyrimidinone, N,N'-Dimethyl propylene urea

251569-25G 251569-100G 251569-500G
220048-302EA 41.14 USD
220048-302 220048-301 220048-303
Tetrahydro-1,3-dimethyl-1H-pyrimidin-2-one
Tetrahydro-1,3-dimethyl-1H-pyrimidin-2-one
Formula: C₆H₁₂N₂O
MW: 128.17 g/mol
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: –24 °C
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 121 °C
Storage Temperature: Ambient
MDL Number: MFCD00006550
CAS Number: 7226-23-5
EINECS: 230-625-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR