Ethyl-3-hydroxybenzoate ≥99%

Synonyms: 3-Hydroxybenzoic acid ethyl ester, Ethyl 3-hydroxybenzoate

251208-100G 251208-25G
220048-276EA 85.08 USD
220048-276 220048-277
Ethyl-3-hydroxybenzoate ≥99%
Ethyl-3-hydroxybenzoate
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 282 °C (1013 hPa)
Melting Pt: 72…73 °C
Density: 1.068 g/cm³ (131 °C)
MDL Number: MFCD00002296
CAS Number: 7781-98-8
EINECS: 231-951-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR