α,α,α,α',α',α'-Hexafluoro-m-xylene ≥99%

Danger

Synonyms: 1,3-Bis(trifluoromethyl)benzene, α,α,α,β,β,β-Hexafluoro-m-xylene

251186-25G 251186-100G
220048-274EA 38.77 USD
220048-274 220048-273
α,α,α,α',α',α'-Hexafluoro-m-xylene ≥99%
α,α,α,α',α',α'-Hexafluoro-m-xylene
Formula: C₈H₄F₆
MW: 214.11 g/mol
Boiling Pt: 116.3 °C (1013 hPa)
Melting Pt: -35 °C
Density: 1.383 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000392
CAS Number: 402-31-3
EINECS: 206-939-4
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR