2-Hydroxynicotinic acid

Synonyms: 2-Hydroxypyridine-3-carboxylic acid, 1,2-Dihydro-2-oxonicotinic acid, 2-Hydroxy-3-pyridinecarboxylic acid, 2-Oxo-1,2-dihydropyridine-3-carboxylic acid

251054-25G 251054-100G
220048-257EA 77.75 USD
220048-257 220048-256
2-Hydroxynicotinic acid
2-Hydroxynicotinic acid
Formula: C₆H₅NO₃
MW: 139.11 g/mol
MDL Number: MFCD00010100
CAS Number: 609-71-2
EINECS: 210-198-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR