2,5-Dibromotoluene ≥98%

Warning

Synonyms: 1,4-Dibromo-2-methylbenzene

250988-5G 250988-25G
220048-240EA 52.77 USD
220048-240 220048-239
2,5-Dibromotoluene ≥98%
2,5-Dibromotoluene
Formula: C₇H₆Br₂
MW: 249.93 g/mol
Boiling Pt: 236 °C (1013 hPa)
Melting Pt: 6 °C
Density: 1.8127 g/cm³ (19 °C)
MDL Number: MFCD00000090
CAS Number: 615-59-8
EINECS: 210-437-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR