2-Ethoxyphenol ≥98%

Warning

Synonyms: 1-Hydroxy-2-ethoxybenzene, Pyrocatechol monoethyl ether, o-Ethoxyphenol

250910-100G 250910-500G
220047-965EA 61.49 USD
220047-965 220047-966
2-Ethoxyphenol ≥98%
2-Ethoxyphenol
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 214 - 216 °C (1013 hPa)
Melting Pt: 28 - 29 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00002187
CAS Number: 94-71-3
EINECS: 202-358-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR