2,4-Difluorophenyl isocyanate ≥99%

Synonyms: 2,4-Difluorophenylisocyanate

250759-25G 250759-5G
220047-951EA 80.77 USD
220047-951 220047-952
2,4-Difluorophenyl isocyanate ≥99%
2,4-Difluorophenyl isocyanate
Formula: C₇H₃F₂NO
MW: 155.1 g/mol
Boiling Pt: 42 °C (6 mmHg)
Density: 1.3 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001997
CAS Number: 59025-55-7
UN: 3080
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR