2-Naphthoyl chloride

Synonyms: 2-Naphthoyl Chloride

250260-10G 250260-50G
220048-189EA 134.62 USD
220048-189 220048-190
2-Naphthoyl chloride
2-Naphthoyl chloride
Formula: C₁₁H₇ClO
MW: 190.63 g/mol
Boiling Pt: 307 °C (1013 hPa)
Melting Pt: 49…52 °C
MDL Number: MFCD00004093
CAS Number: 2243-83-6
EINECS: 218-822-5
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR