Methyl red ACS

Synonyms: 2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid

250198-100G 250198-25G
220048-168EA 348.6 USD
220048-168 220048-169
Methyl red ACS
Methyl red
Formula: C₁₅H₁₅N₃O₂
MW: 269.3 g/mol
Melting Pt: 178…182 °C
Density: 1.31 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002425
CAS Number: 493-52-7
EINECS: 207-776-1
Merck Index: 12,06199

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR